Stampa…

Anti-tRNA Synthetase, Alanyl Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8663-70CEA 825 CHF
USBIT8663-70C
Anti-tRNA Synthetase, Alanyl Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: tRNA Synthetase, Alanyl
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR