Stampa…

Anti-tRNA Synthetase, Alanyl Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT8663-70BEA 774 CHF
USBIT8663-70B
Anti-tRNA Synthetase, Alanyl Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1: 32000; Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: tRNA Synthetase, Alanyl
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR