Stampa…

Anti-NTRK2 Rabbit Monoclonal Antibody [clone: 7H78]

Fornitore: United States Biological
USBIT8663-20CEA 739 CHF
USBIT8663-20C
Anti-NTRK2 Rabbit Monoclonal Antibody [clone: 7H78]
Anticorpi
Recommended dilution: Flow Cytometry: 1:200; Immunochemistry: 1:80

Type: Primary
Antigen: NTRK2 (neurotrophic tyrosine kinase, receptor, type 2)
Clonality: monoclonal
Clone: 7H78
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR