Stampa…

Anti-Triticum vulgare Goat Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT8653-02BEA 521 CHF
USBIT8653-02B
Anti-Triticum vulgare Goat Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: Triticum vulgare
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR