Stampa…

Anti-TPP1 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT8570-01EA 774 CHF
USBIT8570-01
Anti-TPP1 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:4000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: TPP1 (tripeptidyl peptidase I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR