Stampa…

Anti-TRAIP Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8495-03BEA 825 CHF
USBIT8495-03B
Anti-TRAIP Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: TRAIP (TRAF interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR