Stampa…
Anti-TPI1 Sheep Polyclonal Antibody
Fornitore: United States Biological
USBIT8489-86AEA 1070 CHF
USBIT8489-86A
Anti-TPI1 Sheep Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TPI1 (triosephosphate isomerase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Visualizza i codici
Learn more
About VWR
Mehr About VWR