Stampa…
Anti-TPI1 Rabbit Polyclonal Antibody (Biotin)
Fornitore: United States Biological
USBIT8489-85BEA 791 CHF
USBIT8489-85B
Anti-TPI1 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: TPI1 (triosephosphate isomerase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici
Learn more
About VWR
Mehr About VWR