Stampa…

Anti-TRIM29 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT8466-29GEA 774 CHF
USBIT8466-29G
Anti-TRIM29 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:128000; Western Blot: 0. 3-1 ?g/ml; Immunochemistry: 1-2?g/ml

Type: Primary
Antigen: TRIM29 (tripartite motif containing 29)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR