Stampa…
Anti-TRIM29 Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT8466-29EEA 712 CHF
USBIT8466-29E
Anti-TRIM29 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:100-1:1000; Immunochemistry: 1:100
Type: Primary
Antigen: TRIM29 (tripartite motif containing 29)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR