Stampa…

Anti-TRIM63 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT8465-63EEA 774 CHF
USBIT8465-63E
Anti-TRIM63 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:32000

Type: Primary
Antigen: TRIM63 (tripartite motif containing 63, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR