Stampa…

Anti-TRAT1 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT8464-35BEA 1100 CHF
USBIT8464-35B
Anti-TRAT1 Goat Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TRAT1 (T cell receptor associated transmembrane adaptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR