Stampa…

Anti-Triiodothyronine Sheep Polyclonal Antibody

Fornitore: United States Biological
USBIT8425-07AEA 767 CHF
USBIT8425-07A
Anti-Triiodothyronine Sheep Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Triiodothyronine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR