Stampa…

Anti-Tetrodotoxin Rabbit Polyclonal Antibody (Unconjugated)

Fornitore: United States Biological
USBIT8415-50EA 1040 CHF
USBIT8415-50
Anti-Tetrodotoxin Rabbit Polyclonal Antibody (Unconjugated)
Anticorpi
Type: Primary
Antigen: Tetrodotoxin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR