Stampa…

Anti-Trichomonas vaginalis Mouse Monoclonal Antibody [clone: 12K235]

Fornitore: United States Biological
USBIT8350-04FEA 606 CHF
USBIT8350-04F
Anti-Trichomonas vaginalis Mouse Monoclonal Antibody [clone: 12K235]
Anticorpi
Recommended dilution: Immunofluorescence: 1:10-1:50; ELISA: 1:20-1:200; Western Blot: 1:10-1:50; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: Trichomonas vaginalis
Clonality: monoclonal
Clone: 12K235
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR