Stampa…

Anti-Trichomonas vaginalis Mouse Monoclonal Antibody [clone: 11H52]

Fornitore: United States Biological
USBIT8350-04EA 380 CHF
USBIT8350-04
Anti-Trichomonas vaginalis Mouse Monoclonal Antibody [clone: 11H52]
Anticorpi
Type: Primary
Antigen: Trichomonas vaginalis
Clonality: monoclonal
Clone: 11H52
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR