Stampa…
Anti-TRF4 Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT8302-50BEA 889 CHF
USBIT8302-50B
Anti-TRF4 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: TRF4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR