Stampa…
Anti-TREX2 Goat Polyclonal Antibody
Fornitore: United States Biological
USBIT8300-90AEA 774 CHF
USBIT8300-90A
Anti-TREX2 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:128000
Type: Primary
Antigen: TREX2 (three prime repair exonuclease 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Mehr About VWR