Stampa…
Anti-Treponema pallidum Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT8300-11EA 589 CHF
USBIT8300-11
Anti-Treponema pallidum Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunofluorescence: 1:3000
Type: Primary
Antigen: Treponema pallidum
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici
Learn more
About VWR
Mehr About VWR