Stampa…

Anti-TREM1 Mouse Monoclonal Antibody (Biotin) [clone: 9F97]

Fornitore: United States Biological
USBIT8282-09AEA 448 CHF
USBIT8282-09A
Anti-TREM1 Mouse Monoclonal Antibody (Biotin) [clone: 9F97]
Anticorpi
Type: Primary
Antigen: TREM1 (triggering receptor expressed on myeloid cells 1)
Clonality: monoclonal
Clone: 9F97
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR