Stampa…

Anti-TREK1 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8280-01EA 797 CHF
USBIT8280-01
Anti-TREK1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: TREK1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR