Stampa…

Anti-Trefoil Factor 3 Mouse Monoclonal Antibody [clone: 3D9]

Fornitore: United States Biological
USBIT8269-75DEA 988 CHF
USBIT8269-75D
Anti-Trefoil Factor 3 Mouse Monoclonal Antibody [clone: 3D9]
Anticorpi
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: Trefoil Factor 3
Clonality: monoclonal
Clone: 3D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR