Stampa…

Anti-TRAP150 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8266-99REA 712 CHF
USBIT8266-99R
Anti-TRAP150 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.1-1 ?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: TRAP150
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR