Stampa…

Anti-TAP1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornitore: United States Biological
USBIT8266-05E-APCEA 1200 CHF
USBIT8266-05E-APC
Anti-TAP1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: TAP1 (transporter 1, ATP-binding cassette, sub-family B (MDR/TAP))
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR