Stampa…

Anti-TMED10 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8264-65EEA 889 CHF
USBIT8264-65E
Anti-TMED10 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TMED10 (transmembrane emp24-like trafficking protein 10 (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR