Stampa…

Anti-TGM7 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT8260-35CEA 774 CHF
USBIT8260-35C
Anti-TGM7 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:16000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: TGM7 (transglutaminase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Pig
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR