Stampa…

Anti-TGM4 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8260-20BEA 825 CHF
USBIT8260-20B
Anti-TGM4 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Flow Cytometry: 1:10-1:50; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: TGM4 (transglutaminase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR