Stampa…

Anti-Transglutaminase Mouse Monoclonal Antibody [clone: 2A1]

Fornitore: United States Biological
USBIT8260-01JEA 889 CHF
USBIT8260-01J
Anti-Transglutaminase Mouse Monoclonal Antibody [clone: 2A1]
Anticorpi
Recommended dilution: Western Blot: 1:2000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: Transglutaminase
Clonality: monoclonal
Clone: 2A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR