Stampa…

Anti-Transglutaminase Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8260-01GEA 825 CHF
USBIT8260-01G
Anti-Transglutaminase Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: Transglutaminase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR