Stampa…
Anti-Transforming Growth Factor beta Induced Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT8252-91EA 739 CHF
USBIT8252-91
Anti-Transforming Growth Factor beta Induced Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50; Immunofluorescence: 1:100
Type: Primary
Antigen: Transforming Growth Factor beta Induced
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR