Stampa…

Anti-Transforming Growth Factor beta Receptor III Goat Polyclonal Antibody (PE (Phycoerythrin))

Fornitore: United States Biological
USBIT8250-50GEA 948 CHF
USBIT8250-50G
Anti-Transforming Growth Factor beta Receptor III Goat Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Do not freeze!

Type: Primary
Antigen: Transforming Growth Factor beta Receptor III
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR