Stampa…

Anti-TGFBR1 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8250-31DEA 948 CHF
USBIT8250-31D
Anti-TGFBR1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 15 ?g/ml

Type: Primary
Antigen: TGFBR1 (transforming growth factor, beta receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR