Stampa…

Anti-Transforming Growth Factor beta 1 Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 10K72]

Fornitore: United States Biological
USBIT8250-16F2EA 756 CHF
USBIT8250-16F2
Anti-Transforming Growth Factor beta 1 Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 10K72]
Anticorpi
Type: Primary
Antigen: Transforming Growth Factor beta 1
Clonality: monoclonal
Clone: 10K72
Conjugation: Carboxyfluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR