Stampa…

Anti-TGFA Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8250-06KEA 891 CHF
USBIT8250-06K
Anti-TGFA Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:800; Immunochemistry: 1:1600

Type: Primary
Antigen: TGFA (transforming growth factor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR