Stampa…
Anti-TF Rabbit Polyclonal Antibody (Biotin)
Fornitore: United States Biological
USBIT8199-13C1EA 1090 CHF
USBIT8199-13C1
Anti-TF Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Recommended dilution: ELISA: 1:15000-1:700000
Type: Primary
Antigen: TF (transferrin)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR