Stampa…

Anti-Transcriptional Regulator, Yeast Chicken Polyclonal Antibody

Fornitore: United States Biological
USBIT8197-65EA 667 CHF
USBIT8197-65
Anti-Transcriptional Regulator, Yeast Chicken Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Transcriptional Regulator, Yeast
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Yeast
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR