Stampa…

Anti-TFAP2A Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fornitore: United States Biological
USBIT8195-52L-PEEA 1200 CHF
USBIT8195-52L-PE
Anti-TFAP2A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: TFAP2A (transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha))
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR