Stampa…

Anti-Transcription Factor AP-2 alpha, beta Mouse Monoclonal Antibody [clone: A6/2/2]

Fornitore: United States Biological
USBIT8195-52B9EA 1220 CHF
USBIT8195-52B9
Anti-Transcription Factor AP-2 alpha, beta Mouse Monoclonal Antibody [clone: A6/2/2]
Anticorpi
Type: Primary
Antigen: Transcription Factor AP-2 alpha, beta
Clonality: monoclonal
Clone: A6/2/2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR