Stampa…

Anti-Transcription Factor 7-like 1 Sheep Polyclonal Antibody

Fornitore: United States Biological
USBIT8195-42BEA 1030 CHF
USBIT8195-42B
Anti-Transcription Factor 7-like 1 Sheep Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.2?g/ml

Type: Primary
Antigen: Transcription Factor 7-like 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR