Stampa…
Anti-TCF7 Rabbit Polyclonal Antibody (Biotin)
Fornitore: United States Biological
USBIT8195-36A-BIOTEA 1200 CHF
USBIT8195-36A-BIOT
Anti-TCF7 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: TCF7 (transcription factor 7 (T-cell specific, HMG-box))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Mehr About VWR