Stampa…

Anti-TCF7 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8195-36AEA 825 CHF
USBIT8195-36A
Anti-TCF7 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: TCF7 (transcription factor 7 (T-cell specific, HMG-box))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR