Stampa…

Anti-TCERG1L Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8195-05EA 825 CHF
USBIT8195-05
Anti-TCERG1L Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: TCERG1L (transcription elongation regulator 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR