Stampa…

Anti-Transaldolase Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT8192-50BEA 791 CHF
USBIT8192-50B
Anti-Transaldolase Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: Transaldolase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR