Stampa…

Anti-TICAM2 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT8190-03EA 1100 CHF
USBIT8190-03
Anti-TICAM2 Goat Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TICAM2 (toll-like receptor adaptor molecule 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR