Stampa…

Anti-TNFRSF10C Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8181-15-100EA 665 CHF
USBIT8181-15-100 USBIT8181-15-50
Anti-TNFRSF10C Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TNFRSF10C (tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR