Stampa…

Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 11n198]

Fornitore: United States Biological
USBIT8181-11CEA 957 CHF
USBIT8181-11C
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 11n198]
Anticorpi
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: TNFRSF10B (tumor necrosis factor receptor superfamily, member 10b)
Clonality: monoclonal
Clone: 11n198
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR