Stampa…

Anti-TNFSF10 Mouse Monoclonal Antibody [clone: 6D808]

Fornitore: United States Biological
USBIT8175-04CEA 907 CHF
USBIT8175-04C
Anti-TNFSF10 Mouse Monoclonal Antibody [clone: 6D808]
Anticorpi
Type: Primary
Antigen: TNFSF10 (tumor necrosis factor (ligand) superfamily, member 10)
Clonality: monoclonal
Clone: 6D808
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR