Stampa…

Anti-TRAF6BP Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8172-65CEA 712 CHF
USBIT8172-65C
Anti-TRAF6BP Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 2-5 ?g/ml

Type: Primary
Antigen: TRAF6BP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR