Stampa…

Anti-TRAF6 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8172-60DEA 632 CHF
USBIT8172-60D
Anti-TRAF6 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: TRAF6 (TNF receptor-associated factor 6, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Hamster, Human, Monkey, Mouse, Pig, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR