Stampa…

Anti-TRAF5 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT8172-50BEA 642 CHF
USBIT8172-50B
Anti-TRAF5 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000-1:5000; Immunoprecipitation: 1:50-1:200

Type: Primary
Antigen: TRAF5 (TNF receptor-associated factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR